Tisk…

2-Chloro-5-(3,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%

H54708.MDEA 1750 CZK
ALFAH54708.MD ALFAH54708.03
2-Chloro-5-(3,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD22125083
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR