Tisk…

2-Chloro-4-(2-chloro-5-nitrophenylcarbamoyl)benzeneboronic acid, 97%

H54872.MDEA 2450 CZK
ALFAH54872.MD ALFAH54872.03
2-Chloro-4-(2-chloro-5-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C13H9BCl2N2O5
Formula Weight: 354.94
MDL No.: MFCD22125098
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR