Tisk…

2-Chloro-4-(3-nitrophenylcarbamoyl)benzeneboronic acid, 97%

H55240.03EA 11720 CZK
ALFAH55240.03 ALFAH55240.MD
2-Chloro-4-(3-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C13H10BClN2O5
Formula Weight: 320.49
MDL No.: MFCD22125128
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR