Tisk…
2-Chloro-4-(4-methyl-3-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Dodavatel: Thermo Scientific
H55527.03EA 10440 CZK
ALFAH55527.03 ALFAH55527.MD
2-Chloro-4-(4-methyl-3-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClN2O5
Formula Weight: 334.52
MDL No.: MFCD22125146