Tisk…

2-Carboxy-5-chlorobenzeneboronic acid, 97%

H52698.03EA 7410 CZK
ALFAH52698.03 ALFAH52698.MD
2-Carboxy-5-chlorobenzeneboronic acid, 97%
Purity: 97%
CAS No.: 874290-67-2
Molecular Formula: C7H6BClO4
Formula Weight: 200.39
Melting Point: 236-238°
MDL No.: MFCD08235091
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR