Tisk…

2-Chloro-4-(2-fluoro-5-methylphenylcarbamoyl)benzeneboronic acid, 97%

H55404.03EA 10760 CZK
ALFAH55404.03 ALFAH55404.MD
2-Chloro-4-(2-fluoro-5-methylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20091218
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR