Tisk…

2-Chloro-4-(5-fluoro-2-methylphenylcarbamoyl)benzeneboronic acid, 97%

H55726.MDEA 2450 CZK
ALFAH55726.MD ALFAH55726.03
2-Chloro-4-(5-fluoro-2-methylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20040323
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR