Tisk…

2-Chloro-5-(4-fluoro-3-nitrophenylcarbamoyl)benzeneboronic acid, 97%

H55156.MDEA 2570 CZK
ALFAH55156.MD ALFAH55156.03
2-Chloro-5-(4-fluoro-3-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C13H9BClFN2O5
Formula Weight: 338.48
MDL No.: MFCD20040291
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR