Tisk…

2-Fluoro-4-(methylcarbamoyl)phenylboronic acid ≥97%

Dodavatel: Thermo Scientific

Varování

H55727.03EA 23200 CZK
H55727.03 H55727.MD
2-Fluoro-4-(methylcarbamoyl)phenylboronic acid ≥97%
2-Fluoro-4-(methylcarbamoyl)phenylboronic acid
Vzorec: C₈H₉BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD11856043
CAS číslo: 874289-23-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR