Tisk…
4-(2,5-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
H55318.03EA 9770 CZK
ALFAH55318.03 ALFAH55318.MD
4-(2,5-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040300