Tisk…

3-Fluoro-4-(3-methylphenylcarbamoyl)benzeneboronic acid, 97%

H54542.03EA 9770 CZK
ALFAH54542.03 ALFAH54542.MD
3-Fluoro-4-(3-methylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
MDL No.: MFCD20265287
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR