Tisk…

3-(2,3-Dimethylphenylcarbamoyl)-4-fluorobenzeneboronic acid, 97%

H54219.03EA 4980 CZK
ALFAH54219.03 ALFAH54219.MD
3-(2,3-Dimethylphenylcarbamoyl)-4-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040157
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR