Tisk…

3-Fluoro-5-ethoxycarbonylbenzeneboronic acid, 97%

H52444.MDEA 1440 CZK
ALFAH52444.MD ALFAH52444.03
3-Fluoro-5-ethoxycarbonylbenzeneboronic acid, 97%
Purity: 97%
CAS No.: 871329-85-0
Molecular Formula: C9H10BFO4
Formula Weight: 211.98
Melting Point: 146-150°
MDL No.: MFCD07363792
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR