Tisk…

2-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 97%

H52900.03EA 9610 CZK
ALFAH52900.03 ALFAH52900.MD
2-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 97%
Purity: 97%
CAS No.: 850568-04-6
Molecular Formula: C8H8BFO4
Formula Weight: 197.96
Melting Point: 210-212°
MDL No.: MFCD06659868
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR