Tisk…

2-Fluoro-5-carboxybenzeneboronic acid, 96%

H53004.MDEA 1150 CZK
ALFAH53004.MD ALFAH53004.03
2-Fluoro-5-carboxybenzeneboronic acid, 96%
Purity: 96%
CAS No.: 874219-59-7
Molecular Formula: C7H6BFO4
Formula Weight: 183.93
Melting Point: 224-226°
MDL No.: MFCD08436065
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR