Tisk…

Benzeneseleninic acid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

SIAL213004-5GEA 2250 CZK
SIAL213004-5G
Benzeneseleninic acid, Sigma-Aldrich®
Benzeneseleninic acid
Vzorec: C₆H₆O₂Se
MDL Number: MFCD00002100
CAS číslo: 6996-92-5
EINECS: 230-271-2
UN: 3283
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR