Tisk…

4-Methoxy-2-(trifluoromethoxy)benzyl bromide ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

H26398.06EA 11230 CZK
H26398.06 H26398.03
4-Methoxy-2-(trifluoromethoxy)benzyl bromide ≥97%
4-Methoxy-2-(trifluoromethoxy)benzyl bromide
Vzorec: C₉H₈BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06660289
CAS číslo: 886502-56-3
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR