Tisk…

4-(Methylthio)benzylamine

Dodavatel: ABCR GMBH

ABCRAB149996.0005EA 0 CZK
ABCRAB149996.0005
4-(Methylthio)benzylamine
4-(Methylthio)benzylamine
Vzorec: C₈H₁₁NS
MW: 153,2500 g/mol
MDL Number: MFCD01310831
CAS číslo: 83171-39-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR