Tisk…

2-Benzyloxy-5-(trifluoromethyl)benzeneboronic acid, 97%

H52918.MDEA 588 CZK
ALFAH52918.MD ALFAH52918.03
2-Benzyloxy-5-(trifluoromethyl)benzeneboronic acid, 97%
Purity: 97%
CAS No.: 612833-41-7
Molecular Formula: C14H12BF3O3
Formula Weight: 296.05
Melting Point: 122-126°
MDL No.: MFCD06409944
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR