Tisk…

4-Isopropyl-m-toluidinium chloride ≥97%

Dodavatel: Thermo Scientific

Nebezpečí

A19754.14EA 10020 CZK
A19754.14 A19754.06
4-Isopropyl-m-toluidinium chloride ≥97%
4-Isopropyl-m-toluidinium chloride
Vzorec: C₁₀H₁₆ClN
MW: 185,70
Bod tání: 222…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00156562
CAS číslo: 4534-11-6
EINECS: 224-876-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR