Tisk…

5-Benzyl(tert-butyl)carbamoyl-2-chlorobenzeneboronic acid, 97%

H54039.MDEA 1230 CZK
ALFAH54039.MD ALFAH54039.03
5-Benzyl(tert-butyl)carbamoyl-2-chlorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C18H21BClNO3
Formula Weight: 345.63
MDL No.: MFCD22125035
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR