Tisk…
5-[Benzyl(ethyl)carbamoyl]-2-chlorobenzeneboronic acid, 97%
H54352.03EA 4890 CZK
ALFAH54352.03 ALFAH54352.MD
5-[Benzyl(ethyl)carbamoyl]-2-chlorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C16H17BClNO3
Formula Weight: 317.58
MDL No.: MFCD22125065