Tisk…

2-Chloro-4-(4-chlorobenzylcarbamoyl)benzeneboronic acid, 97%

H54479.MDEA 1530 CZK
ALFAH54479.MD ALFAH54479.03
2-Chloro-4-(4-chlorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BCl2NO3
Formula Weight: 323.97
MDL No.: MFCD22125072
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR