Tisk…

2-Chloro-4-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%

H55924.MDEA 2040 CZK
ALFAH55924.MD ALFAH55924.03
2-Chloro-4-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD22125184
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR