Tisk…

4-Benzylcarbamoyl-2-chlorobenzeneboronic acid, 97%

H54223.MDEA 2450 CZK
ALFAH54223.MD ALFAH54223.03
4-Benzylcarbamoyl-2-chlorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H13BClNO3
Formula Weight: 289.52
MDL No.: MFCD22125049
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR