Tisk…

3-Chloro-4-(benzylcarbamoyl)benzeneboronic acid, 98%

H53334.03EA 4190 CZK
ALFAH53334.03 ALFAH53334.MD
3-Chloro-4-(benzylcarbamoyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 850589-42-3
Molecular Formula: C14H13BClNO3
Formula Weight: 289.53
Melting Point: 126-128°
MDL No.: MFCD07363769
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR