Tisk…

4-Chloro-3-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%

H54471.MDEA 939 CZK
ALFAH54471.MD ALFAH54471.03
4-Chloro-3-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD20265275
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR