Tisk…

2-Chloro-4-(4-methoxybenzylcarbamoyl)benzeneboronic acid, 97%

H55200.MDEA 1750 CZK
ALFAH55200.MD ALFAH55200.03
2-Chloro-4-(4-methoxybenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO4
Formula Weight: 319.55
MDL No.: MFCD22125123
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR