Tisk…

3-Chloro-4-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

H54300.MDEA 1120 CZK
ALFAH54300.MD ALFAH54300.03
3-Chloro-4-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20040168
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR