Tisk…

2-Chloro-5-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

H55631.03EA 7000 CZK
ALFAH55631.03 ALFAH55631.MD
2-Chloro-5-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClFNO3
Formula Weight: 307.51
MDL No.: MFCD20040318
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR