Tisk…

4-[Benzyl(methyl)carbamoyl]-3-fluorobenzeneboronic acid, 97%

H54553.MDEA 2450 CZK
ALFAH54553.MD ALFAH54553.03
4-[Benzyl(methyl)carbamoyl]-3-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.10
MDL No.: MFCD20040219
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR