Tisk…

4-Benzylcarbamoyl-3-fluorobenzeneboronic acid, 97%

H52982.MDEA 1650 CZK
ALFAH52982.MD ALFAH52982.03
4-Benzylcarbamoyl-3-fluorobenzeneboronic acid, 97%
Purity: 97%
CAS No.: 874289-19-7
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
Melting Point: 113-116°
MDL No.: MFCD08689501
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR