Tisk…

4-Fluoro-3-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 98%

H54878.03EA 1780 CZK
ALFAH54878.03 ALFAH54878.MD
4-Fluoro-3-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 98%
Purity: 98%
Molecular Formula: C14H12BF2NO3
Formula Weight: 291.06
MDL No.: MFCD20040263
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR