Tisk…
4-Fluoro-3-(3-methylbenzylcarbamoyl)benzeneboronic acid, 98%
H54365.03EA 977 CZK
ALFAH54365.03 ALFAH54365.MD
4-Fluoro-3-(3-methylbenzylcarbamoyl)benzeneboronic acid, 98%
Purity: 98%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.10
MDL No.: MFCD20265251