Tisk…

4-Fluoro-3-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 98%

H54508.MDEA 2450 CZK
ALFAH54508.MD ALFAH54508.03
4-Fluoro-3-(4-fluorobenzylcarbamoyl)benzeneboronic acid, 98%
Purity: 98%
Molecular Formula: C14H12BF2NO3
Formula Weight: 291.06
MDL No.: MFCD20040207
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR