Tisk…
3-Fluoro-5-(3-methylbenzylcarbamoyl)benzeneboronic acid, 97%
H54330.MDEA 2450 CZK
ALFAH54330.MD ALFAH54330.03
3-Fluoro-5-(3-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.10
MDL No.: MFCD20265244