Tisk…

3-Fluoro-5-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%

H54854.03EA 9770 CZK
ALFAH54854.03 ALFAH54854.MD
3-Fluoro-5-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.10
MDL No.: MFCD20265357
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR