Tisk…

5-Benzylcarbamoyl-2-fluorobenzeneboronic acid, 97%

H52684.MDEA 1720 CZK
ALFAH52684.MD ALFAH52684.03
5-Benzylcarbamoyl-2-fluorobenzeneboronic acid, 97%
Purity: 97%
CAS No.: 874289-53-9
Molecular Formula: C14H13BFNO3
Formula Weight: 273.07
Melting Point: 224-226°
MDL No.: MFCD08436031
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR