Tisk…

3-Fluoro-4-(4-methoxybenzyloxycarbamoyl)benzeneboronic acid, 98%

H53049.03EA 4150 CZK
ALFAH53049.03 ALFAH53049.MD
3-Fluoro-4-(4-methoxybenzyloxycarbamoyl)benzeneboronic acid, 98%
Purity: 98%
CAS No.: 913835-49-1
Molecular Formula: C15H15BFNO5
Formula Weight: 319.10
Melting Point: 150-152°
MDL No.: MFCD08235097
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR