Tisk…

2-Amino-4-phenylphenol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL394114-1GEA 1690 CZK
SIAL394114-1G
2-Amino-4-phenylphenol, Sigma-Aldrich®
2-Amino-4-phenylphenol
Vzorec: C₁₂H₁₁NO
MW: 185,2200 g/mol
MDL Number: MFCD00059187
CAS číslo: 1134-36-7
EINECS: 214-484-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR