Tisk…

4-Amino-2,6-diphenylphenol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL273066-1GEA 4310 CZK
SIAL273066-1G
4-Amino-2,6-diphenylphenol, Sigma-Aldrich®
4-Amino-2,6-diphenylphenol
Vzorec: C₁₈H₁₅NO
MDL Number: MFCD00034070
CAS číslo: 50432-01-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR