Tisk…

Isoquinolin-N-oxide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL192694-5GEA 2250 CZK
SIAL192694-5G
Isoquinolin-N-oxide, Sigma-Aldrich®
Isoquinolin-N-oxide
Vzorec: C₉H₇NO
MW: 145,16 g/mol
MDL Number: MFCD00006903
CAS číslo: 1532-72-5
EINECS: 216-242-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR