Tisk…

2-Chlorothioxanthone, Sigma-Aldrich®

Dodavatel: Merck

SIALC72404-5GEA 0 CZK
SIALC72404-5G
2-Chlorothioxanthone, Sigma-Aldrich®
2-Chlorothioxanthone
Vzorec: C₁₃H₇ClOS
MW: 246,72
MDL Number: MFCD00005067
CAS číslo: 86-39-5
EINECS: 201-667-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR