Tisk…

Acetic anhydride-[¹³C₄,²H₆] (99% ¹³C , 97% D), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL607428-100MGEA 28660 CZK
SIAL607428-100MG
Acetic anhydride-[¹³C₄,²H₆] (99% ¹³C , 97% D), Sigma-Aldrich®
Acetic anhydride-[¹³C₄,²H₆]
Vzorec: C₄H₆O₃
MDL Number: MFCD00190254
CAS číslo: 285977-77-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR