Tisk…

Acetonitrile-[¹⁵N] (98% ¹⁵N), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL487864-500MGEA 25420 CZK
SIAL487864-500MG
Acetonitrile-[¹⁵N] (98% ¹⁵N), Sigma-Aldrich®
Acetonitrile-[¹⁵N]
Vzorec: C₂H₃N
MDL Number: MFCD00084189
CAS číslo: 14149-39-4
UN: 1648
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR