Tisk…

1-Nonanesulphonic acid sodium salt, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

74316-10G-FEA 10080 CZK
74316-10G-F
1-Nonanesulphonic acid sodium salt, Supelco®
1-Nonanesulphonic acid sodium salt
Vzorec: C₉H₁₉NaO₃S
MW: 230,30 g/mol
MDL Number: MFCD00025010
CAS číslo: 35192-74-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR