Tisk…

4-Propylphenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL521507-10GEA 0 CZK
SIAL521507-10G
4-Propylphenylboronic acid, Sigma-Aldrich®
4-Propylphenylboronic acid
Vzorec: C₉H₁₃BO₂
MW: 164,01
MDL Number: MFCD00859261
CAS číslo: 134150-01-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR