Tisk…

3-tert-Butyl-5-methylbenzeneboronic acid, 96%

H53352.MDEA 3080 CZK
ALFAH53352.MD
3-tert-Butyl-5-methylbenzeneboronic acid, 96%
Purity: 96%
CAS No.: 193905-93-0
Molecular Formula: C11H17BO2
Formula Weight: 192.06
Melting Point: 158-160°
MDL No.: MFCD03411938
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR