Tisk…

4,6-Dichloro-o-toluidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL569240-5GEA 2260 CZK
SIAL569240-5G
4,6-Dichloro-o-toluidine, Sigma-Aldrich®
4,6-Dichloro-o-toluidine
Vzorec: C₇H₇Cl₂N
MW: 176,05 g/mol
MDL Number: MFCD00044074
CAS číslo: 30273-00-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR