Tisk…

2-Fluoro-5-methylbenzeneboronic acid, 95%

H52714.06EA 4730 CZK
ALFAH52714.06 ALFAH52714.03
2-Fluoro-5-methylbenzeneboronic acid, 95%
Purity: 95%
CAS No.: 166328-16-1
Molecular Formula: C7H8BFO2
Formula Weight: 153.95
Melting Point: 77-82°
MDL No.: MFCD03428512
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR